Order Entry
Denmark
ContactUsLinkComponent
UHPLC columns, Hypersil GOLD™ Vanquish™ aQ
UHPLC columns, Hypersil GOLD™ Vanquish™ aQ
Certificates
UHPLC columns, Hypersil GOLD™ Vanquish™ aQ
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 555-7920
  • 555-7975
  • 555-7974
  • Carbon load:
  • 12%
  • 12%
  • 12%
  • Endcapped:
  • Polar
  • Polar
  • Polar
  • Max. pressure:
  • 1250 bar
  • 1250 bar
  • 1250 bar
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Surface area:
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • Temperature limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • USP code:
  • L1
  • L1
  • L1