Order Entry
Denmark
ContactUsLinkComponent
Anti-EMSY Rabbit Polyclonal Antibody
Anti-EMSY Rabbit Polyclonal Antibody
Catalog # BOSSBS-13069R
Supplier:  Bioss
Anti-EMSY Rabbit Polyclonal Antibody
Catalog # BOSSBS-13069R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    EMSY
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Antigen synonyms:
    Chromosome 11 open reading frame 30|FLJ90741|C11orf30|EMSY_HUMAN.|EMSY protein|GL002|Protein EMSY
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

EMSY binds BRCA2 within a region (exon 3) deleted in cancer. EMSY is capable of silencing the activation potential of BRCA2 exon 3, associates with chromatin regulators HP1 and BS69, and localizes to sites of repair following DNA damage. Its levels are amplified in breast and ovarian cancer.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: EMSY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat