Anti-Surfactant Protein D Antibody
BSBTPB9617
- Antibody type:Primary
- Antigen name:Surfactant Protein D
- Antigen symbol:SFTPD
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6441
- Antigen synonyms:COLEC7|SFTP4|SP-D|PSP-D
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Pulmonary surfactant-associated protein D(SFTPD) detection. Tested with WB, IHC-P, ELISA in Human; Rat.
Type: Primary
Antigen: Surfactant Protein D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat