Order Entry
Denmark
ContactUsLinkComponent
 
Anti-DUSP22 Polyclonal Antibody Pair
  ABNOH00056940-PW1
 :  Abnova
Anti-DUSP22 Polyclonal Antibody Pair
  ABNOH00056940-PW1
 :  Abnova

 

  • Antigen name:
    Dual specificity phosphatase 22
  • Antigen symbol:
    DUSP22
  • Clonality:
    Polyclonal
  • Host:
    Multiple
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    56940
  • Antigen synonyms:
    JSP1|LMWDSP2|MKPX|LMW-DSP2|JKAP|MKP-x|JSP-1|VHX
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-DUSP22 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-DUSP22 (50 µg)

Type:
Antigen: DUSP22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human