Anti-CDC42 + MAPK9 Antibody Pair
Catalog # ABNODI0608
Supplier: Abnova
Specifications
- Antigen symbol:CDC42 + MAPK9
- Host:Multiple
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:998|5601
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CDC42 protein, and the other against the MAPK9 protein for use in Proximity Ligation Assay
Antibody pair set content: 1. CDC42 Rabbit purified Polyclonal Antibody (20 µg) 2. MAPK9 Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: CDC42 + MAPK9
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: