Anti-MPP1 Antibody Pair
Catalog # ABNOH00004354-AP11
Supplier: Abnova
Specifications
- Antigen name:Membrane Palmitoylated Protein 1
- Antigen symbol:MPP1
- Host:Multiple
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:4354
- Antigen synonyms:DXS552E|AAG12|MRG1|55 kDa erythrocyte membrane protein|PEMP|EMP55
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human MPP1.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-MPP1 (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-MPP1, IgG1 kappa (20 µg);
Type:
Antigen: MPP1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human