Order Entry
Denmark
Orders LinkContactUsLinkComponent
Anti-MAP2 Protein Chicken Polyclonal Antibody

Anti-MAP2 Protein Chicken Polyclonal Antibody

50 µl
Catalog # BSENC-1382-50
Supplier:   Biosensis
Anti-MAP2 Protein Chicken Polyclonal Antibody
Catalog # BSENC-1382-50
Supplier:  Biosensis
Anti-MAP2 Protein Chicken Polyclonal Antibody
Anti-MAP2 Protein Chicken Polyclonal Antibody

50 µl

Price
2.450,00 kr
Log in for availability
1 * 50 µl
Product Details & Documents

Microtubules are 25nm diameter protein rods found in most kinds of eukarytic cells. They are polymerized from a dimeric subunit made of one a subunit and one b tubulin subunit. Microtubules are associated with a family of proteins called microtubule associated proteins (MAPs), which includes the protein t (tau) and a group of proteins referred to as MAP1, MAP2, MAP3, MAP4 and MAP5. MAP2 is made up of two ~280kDa apparent molecular weight bands referred to as MAP2a and MAP2b. A third lower molecular weight form, usually called MAP2c, corresponds to a pair of protein bands running at ~70kDa on SDS-PAGE gels. All these MAP2 forms are derived from a single gene by alternate transcription, and all share a C-terminal sequence which includes either three or four microtubule binding peptide sequences, which are very similar to those found in the related microtubule binding protein t (tau). MAP2 isoforms are expressed only in neuronal cells and specifically in the perikarya and dendrites of these cells. Antibodies to MAP2 are therefore excellent markers on neuronal cells, their perikarya and neuronal dendrites.

The antibody reacts with MAP2 protein from a variety of species including human, horse, cow, pig, chicken, rat and mouse. Specificity has been determined by western blot and immunocytochemistry
Application Information:
Immunocytochemistry (IC) at suggested dilution of 1:2,500-5,000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MAP2 Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse, Bovine, Chicken, Horse

Specifications
  • Size:
    50 µl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Antibody type:
    Primary
  • Epitope:
    Multiple points of reactivity likely because of the polyclonal nature of the antibody
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgY
  • Host:
    Chicken
  • Cross adsorption:
    No
  • Antigen name:
    Microtubule-associated protein 2
  • Cat. No.:
    BSENC-1382-50
  • Reactivity:
    Horse|Chicken|Human|Bovine|Rat|Mouse
  • Format:
    Lyophilized
  • Antigen synonyms:
    MAP2 Protein
  • Immunogen:
    Purified full length bovine MAP2 protein
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MAP2
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.