Order Entry
Denmark
ContactUsLinkComponent
Anti-HLA-DQB1 Mouse Polyclonal Antibody
Anti-HLA-DQB1 Mouse Polyclonal Antibody
Catalog # ABNOH00003119-B01
Supplier:  Abnova
Anti-HLA-DQB1 Mouse Polyclonal Antibody
Catalog # ABNOH00003119-B01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Major histocompatibility complex, class II, DQ beta 1
  • Antigen symbol:
    HLA-DQB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3119
  • Antigen synonyms:
    CELIAC1|HLA-DQB|IDDM1
  • Amino acid number:
    1 to 261
  • Storage buffer:
    No additive
  • Sequence:
    MSWKKALRIPGGLRVATVTLMLAMLSTPVAEGRDSPEDFVYQFKGMCYFTNGTERVRLVTRYIYNREEYARFDSDVGVYRAVTPLGPPAAEYWNSQKEVLERTRAELDTVCRHNYQLELRTTLQRRVEPTVTISPSRTEALNHHNLLVCSVTDFYPAQIKVRWFRNDQEETTGVVSTPLIRNGDWTFQILVMLEMTPQRGDVYTCHVEHPSLQNPIIVEWRAQSESAQSKMLSGIGGFVLGLIFLGLGLIIHHRSQKGLLH
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    HLA-DQB1 (AAH12106, 1 a.a. ~ 261 a.a) full-length human protein.
  • Size:
    50 μl
  • Pk:
    50 µl

Specifications

About this item

Mouse polyclonal antibody raised against a full-length human HLA-DQB1 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: HLA-DQB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human