Anti-PPP1R12A Mouse Monoclonal Antibody [clone: AF-20]
ABNOMAB0001
: Abnova
- Antibody type:Primary
- Antigen name:Protein Phosphatase 1, regulatory subunit 12A
- Antigen symbol:PPP1R12A
- Clonality:Monoclonal
- Clone:AF-20
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:4659
- Antigen synonyms:myosin phosphatase-targeting subunit 1|myosin binding subunit|target subunit 1|myosin phosphatase|MYPT1|protein phosphatase 1|M130|protein phosphatase 1 regulatory subunit 12A|regulatory (inhibitor) subunit 12A|MBS|protein phosphatase myosin-binding subunit
- Modification:phospho-Thr696
- Storage buffer:10 mM HEPES/KOH, 150 mM NaCl, pH 7,5 (50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T696 of human MBS/MYPT1.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic phosphopeptide of MBS/MYPT1.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; ELISA: 1 ?g/ml
Type: Primary
Antigen: PPP1R12A
Clonality: Monoclonal
Clone: AF-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat