Anti-CA 19-9 Monoclonal Antibody [clone: 121 SLE]
ABNOMAB3992
: Abnova
- Antibody type:Primary
- Antigen symbol:CA 19-9
- Clonality:Monoclonal
- Clone:121 SLE
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:phosphate buffer, pH 7,3 (0,09% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Immunodiffusion precipitate of ovarian cystic mucins with NS19-9-antibody.
- Size:1 mL
- Pk:1 ml
Mouse monoclonal antibody raised against CA 19-9.
Recommended Dilutions: Immunohistochemistry: 1:10-1:50
Type: Primary
Antigen: CA 19-9
Clonality: Monoclonal
Clone: 121 SLE
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgM
Reactivity: Human