Order Entry
Denmark
ContactUsLinkComponent
Anti-PMS2 Mouse Monoclonal Antibody [clone: 349.29.5.2]
Anti-PMS2 Mouse Monoclonal Antibody [clone: 349.29.5.2]
Catalog # ABNOMAB2089
Supplier:  Abnova
Anti-PMS2 Mouse Monoclonal Antibody [clone: 349.29.5.2]
Catalog # ABNOMAB2089
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Postmeiotic Segregation Increased 2
  • Antigen symbol:
    PMS2
  • Clonality:
    Monoclonal
  • Clone:
    349.29.5.2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Chimpanzee,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5395
  • Antigen synonyms:
    PMS2CL|HNPCC4|mismatch repair endonuclease PMS2|DNA mismatch repair protein PMS2|H_DJ0042M02.9|PMS1 protein homolog 2|PMSL2
  • Storage buffer:
    20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to the first 133 amino acids of human PMS2.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant PMS2.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: PMS2
Clonality: Monoclonal
Clone: 349.29.5.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human, Mouse, Rat