Order Entry
Denmark
ContactUsLinkComponent
 
Anti-BCL3 Hamster Monoclonal Antibody [clone: 150-3.5]
Catalog # ABNOMAB2192
Supplier:  Abnova
Anti-BCL3 Hamster Monoclonal Antibody [clone: 150-3.5]
Catalog # ABNOMAB2192
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B-cell lymphoma 3-encoded protein
  • Antigen symbol:
    BCL3
  • Clonality:
    Monoclonal
  • Clone:
    150-3.5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Hamster
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    12051
  • Antigen synonyms:
    Proto-oncogene BCL3|BCL-3|D19S37|B-cell lymphoma 3 protein|BCL3|BCL4
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant His fusion protein corresponding to mouse Bcl3.
  • Pk:
    100 µG

Specifications

About this item

Hamster monoclonal antibody raised against recombinant Bcl3.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: BCL3
Clonality: Monoclonal
Clone: 150-3.5
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse