Anti-CNXD Monoclonal Antibody [clone: 2E9]
ABNOMAB6587
: Abnova
- Antibody type:Primary
- Antigen name:Calnexin Delta
- Antigen symbol:CNXD
- Clonality:Monoclonal
- Clone:2E9
- Conjugation:Unconjugated
- Host:
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:185-520 aa
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1% proclin, 2% Block Ace)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to Calnexin-delta 185-520.
- Size:50 μg
- Pk:50 µg
Mouse monoclonal antibody raised against synthetic peptide of Calnexin Δ185-520.
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: CNXD
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: 185-520 aa
Host:
Isotype:
Reactivity: