Anti-TOM1L2 Goat Polyclonal Antibody
ABNOPAB6707
: Abnova
- Antibody type:Primary
- Antigen name:Target of Myb1-like 2
- Antigen symbol:TOM1L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:146691
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:CRKKPERSEDALFAL
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human TOM1L2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of TOM1L2.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.1-1 ?g/ml
Type: Primary
Antigen: TOM1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human