Anti-CAD Rabbit Polyclonal Antibody
ABNOPAB17323
: Abnova
- Antibody type:Primary
- Antigen name:Carbamoyl-phosphate Synthetase 2, Aspartate transcarbamylase, And Dihydroorotase
- Antigen symbol:CAD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:790
- Antigen synonyms:CAD trifunctional protein|CAD protein|multifunctional protein CAD
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human CAD.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of CAD.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:20000
Type: Primary
Antigen: CAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse