Order Entry
Denmark
ContactUsLinkComponent
Anti-IgG Goat Monoclonal Antibody (Texas Red®)
Anti-IgG Goat Monoclonal Antibody (Texas Red®)
Catalog # ABNOPAB10619
Supplier:  Abnova
Anti-IgG Goat Monoclonal Antibody (Texas Red®)
Catalog # ABNOPAB10619
Supplier:  Abnova

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Monoclonal
  • Conjugation:
    Texas Red®
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Guinea pig
  • Environmentally Preferable:
  • Epitope:
    F(ab')2
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Form:
    Lyophilized
  • Storage buffer:
    Lyophilized from 20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
  • Storage temperature:
    Store at 4 °C on dry atmosphere. After reconstitution with 1 mL of deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Guinea pig IgG F(ab')2 fragment
  • Purification:
    This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Goat anti-guinea pig IgG recognizes Guinea pig IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host:
Isotype:
Reactivity: Guinea Pig