Anti-CAT Goat Polyclonal Antibody
Catalog # ABNOPAB7048
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Catalase
- Antigen symbol:CAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:847
- Antigen synonyms:Chloramphenicol Acetyltransferase
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DKYNAEKPKNAIHT
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human CAT.
- Purification:Antigen affinity purification
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of CAT.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml
Type: Primary
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat