The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.
The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.
Type: Primary
Antigen: MHC Class II (I-A/I-E)
Clonality:
Clone: M5/114.15.2
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
- Catalog No:
- 80-5321-U100
- 80-5321-U025
- Antigen symbol:
- MHC2
- MHC2
- Antigen name:
- MHC Class 2
- MHC Class 2
- Antigen synonyms:
- MHC II|IA/IE|IA|IE|I-A/E
- MHC II|IA/IE|IA|IE|I-A/E
- Antibody type:
- Primary
- Primary
- Conjugation:
- redFluor® 710
- redFluor® 710
- Clone:
- M5/114.15.2
- M5/114.15.2
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 14961/14969
- 14961/14969
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Tested applications:
- Flow Cytometry
- Flow Cytometry