Anti-VSX2 Rabbit Polyclonal Antibody
ABGEAP16493B
: Abgent
- Antibody type:Primary
- Antigen name:visual system homeobox 2
- Antigen symbol:VSX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Gene ID:338917
- Antigen synonyms:HOX10|MCOPCB3|CHX10|MCOP2|RET1
- Amino acid number:263 to 291
- Storage buffer:pH 7,4
- Molecular weight:39.411 kDa
- Sequence:PKLDKMEQDERGPDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.41
- Shipping temperature:4 °C
- Immunogen:This VSX2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 263-291 amino acids from the C-terminal region of human VSX2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: VSX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human