Order Entry
Denmark
ContactUsLinkComponent
Anti-GOLGA6L9 Rabbit Polyclonal Antibody
Anti-GOLGA6L9 Rabbit Polyclonal Antibody
  ABGEAP18553A
 :  Abgent
Anti-GOLGA6L9 Rabbit Polyclonal Antibody
  ABGEAP18553A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Golgin A6 family-like 9
  • Antigen symbol:
    GOLGA6L9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Gene ID:
    440295
  • Antigen synonyms:
    GOLGA6L20
  • Amino acid number:
    15 to 41
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    49.096 kDa
  • Sequence:
    KAYWQRKSPGIPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.53
  • Shipping temperature:
    4 °C
  • Immunogen:
    This GOLGA6L9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 15-41 amino acids from the N-terminal region of human GOLGA6L9.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: GOLGA6L9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human