- Antibody type:Tag
- Antigen name:Epitope Tag
- Antigen symbol:6xHis
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Size:1 KIT
- Pk:1 KIT
Epitope Tag kit contains:
6X HIS Antibody 100 µg
DYKDDDDK Antibody (Binds to same epitope as Sigma's Anti-FLAG® M2 Antibody)250 µg
GFP Antibody100 µg
GST Antibody 1.0mg
Anti-RABBIT IgG (H&L) (GOAT) Antibody Peroxidase Conjugated 100 µg
Anti-GOAT IgG (H&L) (RABBIT) Antibody Peroxidase Conjugated 100 µg
- Known Cross Reactivity: 6X HIS, FLAG, GFP, GST tagged proteins
Epitope tags are useful for the labeling and detection of proteins using western blotting, immunoprecipitation, and immunostaining techniques. Epitope Tag Antibody Sampler Kit detects overexpressed fusion proteins containing the corresponding epitope tags: FLAG™ tag (DYKDDDDK Tag Antibody) recognizes the DYKDDDDK peptide recognized by Sigma's Anti-FLAG® antibodies, 6X HIS antibody (His-tag or hexahistidine tag), GFP Antibody, and GST Antibody.
6X HIS antibody, GFP Antibody, DYKDDDDK Antibody (Binds to same epitope as Sigma's Anti-FLAG® M2 Antibody), GST Antibody and Conjugated Secondary Antibodies are all affinity purified.
Type: Tag
Antigen: Epitope Tag
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: