Anti-VAMP1 / VAMP2 Mouse Monoclonal Antibody (Unconjugated) [clone: SP10]
Catalog # ROCK200-301-E34
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Vesicle-Associated Membrane Protein 1
- Antigen symbol:VAMP1
- Clonality:Monoclonal
- Clone:SP10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Hamster,Pig,Rabbit,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:6843
- Antigen synonyms:SYB1|SPAX1|VAMP-1
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Synaptobrevin (VAMP) Monoclonal Antibody was produced in mouse by repeated immunizations with synaptic immunoprecipitate (crude) from human brain.
- Purification:purified by chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-Synaptobrevin antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 16 kDa in size corresponding to the VAMP Proteins in Western blot of Rat brain cell lysate.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: VAMP1 / VAMP2
Clonality: Monoclonal
Clone: SP10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Bovine, Mouse, Pig, Rat, Rabbit, Hamster