Anti-UBE2J2 Rabbit Polyclonal Antibody
ROCK600-401-B43S
- Antibody type:Primary
- Antigen name:Ubiquitin-conjugating Enzyme E2, J2
- Antigen symbol:UBE2J2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:118424
- Antigen synonyms:PRO2121|NCUBE2|NCUBE-2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of the human Ube2j2 Protein.
- Purification:affinity-purified
- Size:25 µl
- Pk:25 µl
This affinity purified antibody has been tested for use in ELISA, western blotting and immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 29 kDa in size corresponding to Ube2j2 Protein.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: UBE2J2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human