Order Entry
Denmark
ContactUsLinkComponent
Anti-PSMD14 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 11C612]
Anti-PSMD14 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 11C612]
Catalog # USBIP9078-61-HRP
Supplier:  US Biological
undefined
Anti-PSMD14 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 11C612]
Catalog # USBIP9078-61-HRP
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome (prosome, macropain) 26S subunit, non-ATPase 14
  • Antigen symbol:
    PSMD14
  • Clonality:
    Monoclonal
  • Clone:
    11C612
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: PSMD14 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 14)
Clonality: monoclonal
Clone: 11C612
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human