Anti-PAK6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIP2488-30F-FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:p21 Protein (Cdc42/Rac)-activated kinase 6
- Antigen symbol:PAK6
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:131 to 146
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunoprecipitation: 1:100
Do not freeze!
Type: Primary
Antigen: PAK6 (p21 protein (Cdc42/Rac)-activated kinase 6)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human