Anti-KIR2DS4 Mouse Monoclonal Antibody [clone: 8H113]
Catalog # USBIK1858-61
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4
- Antigen symbol:KIR2DS4
- Clonality:Monoclonal
- Clone:8H113
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: KIR2DS4 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4)
Clonality: monoclonal
Clone: 8H113
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human