Anti-IP3 Receptor Rabbit Polyclonal Antibody
USBII8471-01
- Antibody type:Primary
- Antigen name:IP3 Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: IP3 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat