Anti-HLA Class 1 Antigen ABC Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10D168]
USBIH6098-39H1
- Antibody type:Primary
- Antigen name:HLA Class 1 Antigen ABC
- Clonality:Monoclonal
- Clone:10D168
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HLA Class 1 Antigen ABC
Clonality: monoclonal
Clone: 10D168
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human