Anti-Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3 Rabbit polyclonal antibody
USBIG8576-88
- Antibody type:Primary
- Antigen name:Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 2.5 ?g/ml; Western Blot: 1:500-1:3000; ELISA: 1:10000-1:50000
Type: Primary
Antigen: Golgi Associated, Gamma Adaptin Ear Containing, ARF Binding Protein 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human