Anti-Glycerol-3-Phosphate Dehydrogenase Sheep Polyclonal Antibody (Biotin)
USBIG8154-73B
- Antibody type:Primary
- Antigen name:Glycerol-3-Phosphate Dehydrogenase
- Antigen symbol:GPDH
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2. No
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulphate precipitation and ion exchange chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Glycerol-3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: