Anti-GSTA1 Mouse Monoclonal Antibody [clone: 2F7]
Catalog # USBIG8135-10K
Supplier: US Biological
Specifications
- Antibody type:Tag
- Antigen name:Glutathione S-transferase alpha 1
- Antigen symbol:GSTA1
- Clonality:Monoclonal
- Clone:2F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 222
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Tag
Antigen: GSTA1 (glutathione S-transferase alpha 1)
Clonality: monoclonal
Clone: 2F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human