Anti-GLUT 5 Rabbit polyclonal antibody
USBIG3900-65N
- Antibody type:Primary
- Antigen symbol:GLUT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Guinea pig,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:Serum
- Form:liquid
- Amino acid number:490 to 502
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: GLUT 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea pig, Mouse, Rat