Anti-GABA Receptor A, alpha 4, Rabbit polyclonal antibody
USBIG1015-20F
- Antibody type:Primary
- Antigen name:GABA A Receptor Alpha 4
- Antigen symbol:GABRA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: GABA Receptor A, alpha 4,
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat