Anti-Eukaryotic Translation Initiation Factor 2, alpha Mouse monoclonal antibody [clone: 7H20]
Catalog # USBIE8949-01A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic Translation Initiation Factor 2 alpha
- Antigen symbol:EIF2A
- Clonality:Monoclonal
- Clone:7H20
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:50
Type: Primary
Antigen: Eukaryotic Translation Initiation Factor 2, alpha
Clonality: monoclonal
Clone: 7H20
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse, Rat