Anti-ITGAL Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6D18]
USBIC2262-33J2
- Antibody type:Primary
- Antigen name:Integrin, Alpha L (Antigen CD11A (P180), Lymphocyte Function-Associated Antigen 1, AlphaPolypeptide)
- Antigen symbol:ITGAL
- Clonality:Monoclonal
- Clone:6D18
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rabbit,Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: Monoclonal
Clone: 6D18
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit