Anti-Carnitine O-palmitoyltransferase 1, Brain Isoform Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIC1385-29X1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Carnitine O-palmitoyltransferase 1, Brain Isoform
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Amino acid number:769 to 798
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Carnitine O-palmitoyltransferase 1, Brain Isoform
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse