Anti-BMP4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIB2553-20H-FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:bone morphogenetic Protein 4
- Antigen symbol:BMP4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human