Order Entry
Denmark
ContactUsLinkComponent
Anti-BBOX1 Mouse monoclonal antibody unconjugated [clone: 6H3]
Anti-BBOX1 Mouse monoclonal antibody unconjugated [clone: 6H3]
  USBIB0001-05
undefined
Anti-BBOX1 Mouse monoclonal antibody unconjugated [clone: 6H3]
  USBIB0001-05

 

  • Antibody type:
    Primary
  • Antigen name:
    butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 1
  • Antigen symbol:
    BBOX1
  • Clonality:
    Monoclonal
  • Clone:
    6H3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Amino acid number:
    278 to 388
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: BBOX1 (butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 1)
Clonality: monoclonal
Clone: 6H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human