Anti-ASIP Rabbit Polyclonal Antibody
USBIA1124-21D
- Antibody type:Primary
- Antigen name:Agouti signaling Protein
- Antigen symbol:ASIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:352 to 374
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: ASIP (agouti signaling protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human