Order Entry
Denmark
ContactUsLinkComponent
Anti-Caldesmon, Smooth Mouse Monoclonal Antibody [clone: CD-7]
Anti-Caldesmon, Smooth Mouse Monoclonal Antibody [clone: CD-7]
Catalog # USBI144258
Supplier:  US Biological
Anti-Caldesmon, Smooth Mouse Monoclonal Antibody [clone: CD-7]
Catalog # USBI144258
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caldesmon, Smooth
  • Clonality:
    Monoclonal
  • Clone:
    CD-7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit,
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    ascites
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Caldesmon, Smooth
Clonality: monoclonal
Clone: CD-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit