Anti-EMP2 Rabbit Polyclonal Antibody
USBI137933
- Antibody type:Primary
- Antigen name:Epithelial membrane protein 2
- Antigen symbol:EMP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1. 25 ?g/ml; Immunochemistry: 4-8?g/ml
Type: Primary
Antigen: EMP2 (epithelial membrane protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human