Anti-Oncofetal Antigen 5T4 Mouse Monoclonal Antibody [clone: 1B6]
Catalog # USBI130721
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Oncofetal Antigen 5T4
- Clonality:Monoclonal
- Clone:1B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:219 to 328
- Storage buffer:PBS, pH 7,2.
- Sequence:SNHFLYLPRDVLAQLPSLRHLDLSNNSLVSLTYVSFRNLTHLESLHLEDNALKVLHNGTLAELQGLPHIRVFLDNNPWVCDCHMADMVTWLKETEVVQGKDRLTCAYPEK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Oncofetal Antigen 5T4
Clonality: monoclonal
Clone: 1B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat