Anti-AEN Mouse Polyclonal Antibody
USBI128606
- Antibody type:Primary
- Antigen name:Apoptosis enhancing nuclease
- Antigen symbol:AEN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 325
- Storage buffer:PBS, pH 7,2.
- Sequence:MVPREAPESAQCLCPSLTIPNAKDVLRKRHKRRSRQHQRFMARKALLQEQGLLSMPPEPGSSPLPTPFGAATATEAASSGKQCLRAGSGSAPCSRRPAPGKASGPLPSKCVAIDCEMVGTGPRGRVSELARCSIVSYHGDVLYDKYIRPEMPIADYRTRWSGITRQHMRKAVPFQVAQKEILKLLKGKVVVGHALHNDFQALKYVHPRSQTRDTTYVPNFLSEPGLHTRARVSLKDLALQLLHKKIQVGQHGHSSVEDATTAMELYRLVEVQWEQQEARSLWTCPEDREPDSSTDMEQYMEDQYWPDDLAHGSRGGAREAQDRRN
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: AEN (apoptosis enhancing nuclease)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human