Anti-CBLC Mouse Monoclonal Antibody [clone: 3F10]
USBI124387
- Antibody type:Primary
- Antigen name:Cbl proto-oncogene C, E3 ubiquitin Protein ligase
- Antigen symbol:CBLC
- Clonality:Monoclonal
- Clone:3F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MALAVAPWGRQWEEARALGRAVRMLQRLEEQCVDPRLSVSPPSLRDLLPRTAQLLREVAHSRRAAGGGGPGGPGGSGDFLLIYLANLEAKSRQVAALLPPRGRRSANDELFRAGSRLRRQLAKLAIIFSHMHAELHALFPGGKYCGHMYQLTKAPAHTFWRESCGARCVLPWAEFESLLGTCHPVEPGCTALALRTTIDLTCSGHVSIFEFDVFTRLFQPWPTLLKNWQLLAVNHPGYMAFLTYDEVQERLQACR
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CBLC (Cbl proto-oncogene C, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 3F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human