Anti-ATP4B Mouse Monoclonal Antibody [clone: 1D10]
USBI123705
- Antibody type:Primary
- Antigen name:ATPase, H+/K+ exchanging, beta polypeptide
- Antigen symbol:ATP4B
- Clonality:Monoclonal
- Clone:1D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:67 to 176
- Storage buffer:PBS, pH 7,2.
- Sequence:DPYTPDYQDQLRSPGVTLRPDVYGEKGLEIVYNVSDNRTWADLTQTLHAFLAGYSPAAQEDSINCTSEQYFFQESFRAPNHTKFSCKFTADMLQNCSGLADPNFGFEEGK
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: ATP4B (ATPase, H+/K+ exchanging, beta polypeptide)
Clonality: monoclonal
Clone: 1D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human