Anti-TFCP2L1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI042763-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor CP2-like 1
- Antigen symbol:TFCP2L1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:221 to 251
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50
Type: Primary
Antigen: TFCP2L1 (transcription factor CP2-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse