Anti-REG3G Rabbit Polyclonal Antibody (Biotin)
USBI040947-BIOTIN
- Antibody type:Primary
- Antigen name:Regenerating islet-derived protein 3 gamma
- Antigen symbol:REG3G
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:89 to 116
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: REG3G (regenerating islet-derived 3 gamma)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse