Anti-PIP5KL1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # USBI040079-HRP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylinositol-4-phosphate 5-kinase-like 1
- Antigen symbol:PIP5KL1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Form:liquid
- Amino acid number:64 to 94
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Type: Primary
Antigen: PIP5KL1 (phosphatidylinositol-4-phosphate 5-kinase-like 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human