Anti-CYB5A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI034403-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cytochrome b5 type A (microsomal)
- Antigen symbol:CYB5A
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Amino acid number:68 to 98
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CYB5A (cytochrome b5 type A (microsomal))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human