Anti-Lambda light chain Mouse Monoclonal Antibody [clone: HP6054]
Catalog # USBI031075
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:lambda light chain
- Clonality:Monoclonal
- Clone:HP6054
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-16000; Immunochemistry: 1:10-50
Type: Primary
Antigen: Lambda light chain
Clonality: Monoclonal
Clone: HP6054
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human